4-Iodophenyl isothiocyanate ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-Iodo-4-isothiocyanatobenzene, 4-Iodophenylisothiocyanate
L12583.09
L12583.04
L12583.09EA
4020
CZK
L12583.09
L12583.04
4-Iodophenyl isothiocyanate ≥97%
4-Iodophenyl isothiocyanate
Purity: 97%
CAS No.: 2059-76-9
Molecular Formula: C7H4INS
Formula Weight: 261.09
Odour: Pungent
Melting Point: 74-78°
Beilstein Reference: 2412143
UN No.: 2923
MDL No.: MFCD00041091
Vzorec:
C₇H₄INS MW: 261,09 g/mol Bod varu: 296 °C (1013 hPa) Bod tání: 74…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041091 CAS číslo: 2059-76-9 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...