4-Iodophenyl isocyanate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-Iodophenylisocyanate
L16000.03
L16000.06
L16000.03EA
1240
CZK
L16000.03
L16000.06
4-Iodophenyl isocyanate ≥97%
4-Iodophenyl isocyanate
Purity: 97%
CAS No.: 15845-62-2
Molecular Formula: C7H4INO
Formula Weight: 245.02
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₇H₄INO Bod varu: 84 °C (5 mmHg) Bod tání: 43…47 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013875 CAS číslo: 15845-62-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...