2-Iodobenzoic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
o-Iodobenzoic acid
M-Clarity™ quality level = MQ100
I7675-100G
I7675-25G
SIALI7675-100GEA
2650
CZK
SIALI7675-100G
SIALI7675-25G
2-Iodobenzoic acid, Sigma-Aldrich®
2-Iodobenzoic acid
Vzorec:
C₇H₅IO₂ MW: 248,02 g/mol Bod varu: 314 °C (1013 hPa) Bod tání: 159…160 °C Hustota: 2,25 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002419 CAS číslo: 88-67-5 EINECS: 201-850-7 Merck Index: 13,05052 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...