Tisk…

Ethyl-4-iodobenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester

M-Clarity™ quality level = MQ100

641979-25G 641979-5G
SIAL641979-25GEA 6430 CZK
SIAL641979-25G SIAL641979-5G
Ethyl-4-iodobenzoate, Sigma-Aldrich®
Ethyl-4-iodobenzoate
Vzorec: C₉H₉IO₂
MW: 276,07 g/mol
Bod varu: 281 °C (1013 hPa)
Hustota: 1,655 g/cm³ (20 °C)
MDL Number: MFCD00017344
CAS číslo: 51934-41-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR