Methyl-2-iodobenzoate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Methyl 2-iodobenzoate, 2-Iodobenzoic acid methyl ester
A13204.09
A13204.18
A13204.09EA
896
CZK
A13204.09
A13204.18
Methyl-2-iodobenzoate ≥98%
Methyl-2-iodobenzoate
Purity: 98%
CAS No.: 610-97-9
Molecular Formula: C8H7IO2
Formula Weight: 262.05
Flash Point:
Vzorec:
C₈H₇IO₂ MW: 262,05 g/mol Bod varu: 274 °C Hustota: 1,730 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00016351 CAS číslo: 610-97-9 EINECS: 210-243-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...