Tisk…

4-Iodobenzoyl chloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: p-Iodobenzoyl chloride

B25388.06 B25388.14 B25388.22
B25388.06EA 1370 CZK
B25388.06 B25388.14 B25388.22
4-Iodobenzoyl chloride ≥98%
4-Iodobenzoyl chloride

Purity: 98% CAS No.: 1711-02-0 Molecular Formula: C7H4ClIO Formula Weight: 266.47 Flash Point: 110°(230°F) Melting Point: 61-65° Boiling Point: 120-121°/1mm Beilstein Reference: 774718 UN No.: 3261 MDL No.: MFCD00001058

Vzorec: C₇H₄ClIO
Bod varu: 120…121 °C (1 mmHg)
Bod tání: 61…65 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001058
CAS číslo: 1711-02-0
EINECS: 216-974-7
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR