Tisk…

3-Iodobenzonitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

529036-10G 529036-1G
SIAL529036-10GEA 7760 CZK
SIAL529036-10G SIAL529036-1G
3-Iodobenzonitrile, Sigma-Aldrich®
3-Iodobenzonitrile
Vzorec: C₇H₄IN
MW: 229,02 g/mol
MDL Number: MFCD00079762
CAS číslo: 69113-59-3
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR