Tisk…

Pentafluorobenzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,3,4,5,6-Pentafluorobenzenesulphonyl chloride, Pentafluorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ200

103764-25G 103764-5G
SIAL103764-25GEA 9810 CZK
SIAL103764-25G SIAL103764-5G
Pentafluorobenzenesulphonyl chloride, Sigma-Aldrich®
Pentafluorobenzenesulphonyl chloride
Vzorec: C₆ClF₅O₂S
MW: 266,58 g/mol
Bod varu: 210 °C (1013 hPa)
Hustota: 1,79 g/cm³ (20 °C)
MDL Number: MFCD00007427
CAS číslo: 832-53-1
EINECS: 212-620-0
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR