Pentafluorobenzenesulphonyl chloride ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2,3,4,5,6-Pentafluorobenzenesulphonyl chloride, Pentafluorobenzene-1-sulphonyl chloride
A17676.03
A17676.06
A17676.14
A17676.03EA
817
CZK
A17676.03
A17676.06
A17676.14
Pentafluorobenzenesulphonyl chloride ≥98%
Pentafluorobenzenesulphonyl chloride
Purity: 98+%
CAS No.: 832-53-1
Molecular Formula: C6ClF5O2S
Formula Weight: 266.57
Flash Point: 90°(194°F)
Refractive Index: 1.4790
Boiling Point: 210-211°
Beilstein Reference: 2142325
UN No.: 3265
MDL No.: MFCD00007427
Vzorec:
C₆ClF₅O₂S MW: 266.58 Bod varu: 210…211 °C Hustota: 1.796 Bod vzplanutí: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007427 CAS číslo: 832-53-1 EINECS: 212-620-0 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...