Tisk…

4-Fluorobenzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Fluorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ200

F6206-100G F6206-25G F6206-5G
SIALF6206-100GEA 5290 CZK
SIALF6206-100G SIALF6206-25G SIALF6206-5G
4-Fluorobenzenesulphonyl chloride, Sigma-Aldrich®
4-Fluorobenzenesulphonyl chloride
Vzorec: C₆H₄ClFO₂S
MW: 194,61 g/mol
Bod varu: 263 °C (1013 hPa)
Bod tání: 36 °C
MDL Number: MFCD00007438
CAS číslo: 349-88-2
EINECS: 206-493-0
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR