Pentafluorothiophenol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
2,3,4,5,6-Pentafluorothiophenol, Pentafluorophenyl mercaptan, Pentafluorobenzenethiol, Mercaptopentafluorobenzene
M-Clarity™ quality level = MQ100
P5654-25G-A
P5654-5G-A
SIALP5654-25G-AEA
14560
CZK
SIALP5654-25G-A
SIALP5654-5G-A
Pentafluorothiophenol, Sigma-Aldrich®
Pentafluorothiophenol
Vzorec:
C₆HF₅S MW: 200.13 Bod varu: 143 Bod tání: –24 °C Hustota: 1.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004828 CAS číslo: 771-62-0 EINECS: 212-236-3 UN: 2920 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...