3,5-Difluorophenol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
197572-10G
197572-1G
SIAL197572-10GEA
9870
CZK
SIAL197572-10G
SIAL197572-1G
3,5-Difluorophenol, Sigma-Aldrich®
3,5-Difluorophenol
Vzorec:
C₆H₄F₂O MW: 130,09 g/mol Bod varu: 65…68 °C (15 mmHg) Bod tání: 55 °C Hustota: 1,566 g/cm³ (20 °C) |
MDL Number:
MFCD00002255 CAS číslo: 2713-34-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...