Tisk…

4-Fluoroveratrole ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A17936.06 A17936.14
A17936.06EA 1510 CZK
A17936.06 A17936.14
4-Fluoroveratrole ≥98%
4-Fluoroveratrole

Purity: 98% CAS No.: 398-62-9 Molecular Formula: C8H9FO2 Formula Weight: 156.16 Flash Point: 91°(195°F) Refractive Index: 1.5100 Boiling Point: 120-123°/45mm Beilstein Reference: 2441600 MDL No.: MFCD00012201

Vzorec: C₈H₉FO₂
Bod varu: 120…123 °C (45 mmHg)
Hustota: 1,171
Bod vzplanutí: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012201
CAS číslo: 398-62-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR