Tisk…

3-(Trifluoromethoxy)thiophenol ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3-(Trifluoromethoxy)benzenethiol, 3-(Trifluoromethoxy)benzene-1-thiol

L19912.03 L19912.06
L19912.03EA 2850 CZK
L19912.03 L19912.06
3-(Trifluoromethoxy)thiophenol ≥98%
3-(Trifluoromethoxy)thiophenol

Purity: 98% CAS No.: 220239-66-7 Molecular Formula: C7H5F3OS Formula Weight: 194.18 Refractive Index: 1.4770

Vzorec: C₇H₅F₃OS
MW: 194,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320799
CAS číslo: 220239-66-7
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR