3-Fluorophenyl isothiocyanate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B25001.06
B25001.14
B25001.06EA
1640
CZK
B25001.06
B25001.14
3-Fluorophenyl isothiocyanate ≥97%
3-Fluorophenyl isothiocyanate
Purity: 97+%
CAS No.: 404-72-8
Molecular Formula: C7H4FNS
Formula Weight: 153.18
Flash Point: 85°(185°F)
Refractive Index: 1.6200
Odour: Pungent
Boiling Point: 227°
Beilstein Reference: 2081172
UN No.: 2922
MDL No.: MFCD00004804
Vzorec:
C₇H₄FNS Bod varu: 227 °C Hustota: 1,270 Bod vzplanutí: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004804 CAS číslo: 404-72-8 EINECS: 206-967-7 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...