Tisk…

4-Fluorophenyl isocyanate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Fluorophenylisocyanate, 1-Fluoro-4-isocyanatobenzene

M-Clarity™ quality level = MQ100

F14335-10G F14335-5G
SIALF14335-10GEA 2230 CZK
SIALF14335-10G SIALF14335-5G
4-Fluorophenyl isocyanate, Sigma-Aldrich®
4-Fluorophenyl isocyanate
Vzorec: C₇H₄FNO
MW: 137.11
Bod varu: 42…47 °C (1 mmHg)
Hustota: 1.206 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002023
CAS číslo: 1195-45-5
EINECS: 214-799-0
UN: 3080
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR