4-Fluoro-3-hydroxybenzoic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
571113-1G
571113-5G
SIAL571113-1GEA
1960
CZK
SIAL571113-1G
SIAL571113-5G
4-Fluoro-3-hydroxybenzoic acid, Sigma-Aldrich®
4-Fluoro-3-hydroxybenzoic acid
Vzorec:
C₇H₅FO₃ MW: 156,11 g/mol |
MDL Number:
MFCD00269641 CAS číslo: 51446-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...