5-Fluorosalicylic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
5-Fluoro-2-hydroxybenzoic acid
B21751.03
B21751.06
B21751.03EA
1860
CZK
B21751.03
B21751.06
5-Fluorosalicylic acid ≥97%
5-Fluorosalicylic acid
Purity: 97%
CAS No.: 345-16-4
Molecular Formula: C7H5FO3
Formula Weight: 156.11
Melting Point: 178-182°
Beilstein Reference: 2209120
Vzorec:
C₇H₅FO₃ MW: 156.11 Bod varu: 295 Bod tání: 178…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002456 CAS číslo: 345-16-4 EINECS: 206-455-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...