2,6-Difluoro-4-hydroxybenzoic acid 95%
Dodavatel: Thermo Scientific
|
Varování
|
H26134.MD
H26134.03
H26134.MDEA
1430
CZK
H26134.MD
H26134.03
2,6-Difluoro-4-hydroxybenzoic acid 95%
2,6-Difluoro-4-hydroxybenzoic acid
Purity: 95%
CAS No.: 214917-68-7
Molecular Formula: C7H4F2O3
Formula Weight: 174.10
Melting Point: 170-171° dec.
MDL No.: MFCD03094500
Vzorec:
C₇H₄F₂O₃ MW: 174,1 g/mol Bod tání: 170…171 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD03094500 CAS číslo: 214917-68-7 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...