3-Fluoro-4-methoxybenzoic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
365114-1G
365114-5G
SIAL365114-1GEA
1350
CZK
SIAL365114-1G
SIAL365114-5G
3-Fluoro-4-methoxybenzoic acid, Sigma-Aldrich®
3-Fluoro-4-methoxybenzoic acid
Vzorec:
C₈H₇FO₃ MW: 170,14 g/mol Bod tání: 211,5 °C |
MDL Number:
MFCD00060675 CAS číslo: 403-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...