Tisk…

2,4-Difluoro-3-methoxybenzoic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B25597.06 B25597.14
B25597.06EA 2400 CZK
B25597.06 B25597.14
2,4-Difluoro-3-methoxybenzoic acid ≥98%
2,4-Difluoro-3-methoxybenzoic acid

Purity: 98+% CAS No.: 178974-97-5 Molecular Formula: C8H6F2O3 Formula Weight: 188.13 Melting Point: 193-195° MDL No.: MFCD02258905

Vzorec: C₈H₆F₂O₃
MW: 188.13
Bod tání: 193…195 °C
Storage Temperature: Ambient
MDL Number: MFCD02258905
CAS číslo: 178974-97-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR