4-Amino-2-fluorobenzoic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
650625-1G
650625-250MG
SIAL650625-1GEA
7030
CZK
SIAL650625-1G
SIAL650625-250MG
4-Amino-2-fluorobenzoic acid, Sigma-Aldrich®
4-Amino-2-fluorobenzoic acid
Vzorec:
C₇H₆FNO₂ MW: 155,13 g/mol |
MDL Number:
MFCD01569397 CAS číslo: 446-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...