5-Fluoroanthranilic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
2-Amino-5-fluorobenzoic acid
M-Clarity™ quality level = MQ100
367982-10G
367982-1G
SIAL367982-10GEA
11400
CZK
SIAL367982-10G
SIAL367982-1G
5-Fluoroanthranilic acid, Sigma-Aldrich®
5-Fluoroanthranilic acid
Vzorec:
C₇H₆FNO₂ MW: 155.13 Bod varu: 305 Bod tání: 186 Storage Temperature: Ambient |
MDL Number:
MFCD00055566 CAS číslo: 446-08-2 EINECS: 207-159-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...