3-Fluorobenzoic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
F6605-25G
F6605-5G
SIALF6605-25GEA
1350
CZK
SIALF6605-25G
SIALF6605-5G
3-Fluorobenzoic acid, Sigma-Aldrich®
3-Fluorobenzoic acid
Vzorec:
C₇H₅FO₂ MW: 140.11 Bod varu: 251 Bod tání: 122 Storage Temperature: Ambient |
MDL Number:
MFCD00002489 CAS číslo: 455-38-9 EINECS: 207-248-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...