2-Fluorobenzhydrazide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Fluorobenzohydrazide, 2-Fluorobenzene-1-carbohydrazide, 2-Fluorobenzoic hydrazide, 2-Fluorobenzoic acid hydrazide
B24921.09
B24921.18
B24921.30
B24921.09EA
1270
CZK
B24921.09
B24921.18
B24921.30
2-Fluorobenzhydrazide ≥98%
2-Fluorobenzhydrazide
Purity: 98%
CAS No.: 446-24-2
Molecular Formula: C7H7FN2O
Formula Weight: 154.14
Melting Point: 72-74°
MDL No.: MFCD00025112
Vzorec:
C₇H₇FN₂O MW: 154,14 g/mol Bod tání: 72…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025112 CAS číslo: 446-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...