Tisk…

Pentafluorobenzamide ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,3,4,5,6-Pentafluorobenzamide

A18397.06 A18397.14 A18397.22
A18397.06EA 1770 CZK
A18397.06 A18397.14 A18397.22
Pentafluorobenzamide ≥99%
Pentafluorobenzamide

Purity: 99% CAS No.: 652-31-3 Molecular Formula: C7H2F5NO Formula Weight: 211.09 Melting Point: 146-149° MDL No.: MFCD00007971

Vzorec: C₇H₂F₅NO
MW: 211.09
Bod varu: 272
Bod tání: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00007971
CAS číslo: 652-31-3
EINECS: 211-488-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR