4-((Trifluoromethyl)thio)benzoyl chloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
555479-1G
555479-5G
SIAL555479-1GEA
2070
CZK
SIAL555479-1G
SIAL555479-5G
4-((Trifluoromethyl)thio)benzoyl chloride, Sigma-Aldrich®
4-((Trifluoromethyl)thio)benzoyl chloride
Vzorec:
C₈H₄ClF₃OS MW: 240,63 g/mol Bod varu: 96 °C (18 mmHg) Hustota: 1,43 g/cm³ (20 °C) |
MDL Number:
MFCD01631632 CAS číslo: 330-14-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...