Tisk…

4-Fluorobenzoyl chloride ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: p-Fluorobenzoyl chloride

A12946.14 A12946.22 A12946.36
A12946.14EA 1130 CZK
A12946.14 A12946.22 A12946.36
4-Fluorobenzoyl chloride ≥98%
4-Fluorobenzoyl chloride

Purity: 98% CAS No.: 403-43-0 Molecular Formula: C7H4ClFO Formula Weight: 158.56 Flash Point: 82°(179°F) Refractive Index: 1.5320 Odour: Acidic Melting Point: 10-12° Boiling Point: 193° Beilstein Reference: 386215 UN No.: 3265 MDL No.: MFCD00000684

Vzorec: C₇H₄ClFO
MW: 158,56 g/mol
Bod varu: 193 °C
Bod tání: 10…12 °C
Hustota: 1,326
Bod vzplanutí: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000684
CAS číslo: 403-43-0
EINECS: 206-961-4
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR