3-Fluorobenzoyl chloride ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-Fluorobenzoylchloride, m-Fluorobenzoyl chloride
L09152.06
L09152.14
L09152.06EA
807
CZK
L09152.06
L09152.14
3-Fluorobenzoyl chloride ≥98%
3-Fluorobenzoyl chloride
Purity: 98%
CAS No.: 1711-07-5
Molecular Formula: C7H4ClFO
Formula Weight: 158.56
Flash Point: 82°(179°F)
Refractive Index: 1.5285
Odour: Pungent
Melting Point: -30°
Boiling Point: 188-189°
Beilstein Reference: 636610
UN No.: 3265
MDL No.: MFCD00000670
Vzorec:
C₇H₄ClFO MW: 158,56 g/mol Bod varu: 188…189 °C Bod tání: –30 °C Hustota: 1,320 Bod vzplanutí: 82 °C (179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000670 CAS číslo: 1711-07-5 EINECS: 216-977-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...