2-Fluorobenzoyl chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
o-Fluorobenzoyl chloride
M-Clarity™ quality level = MQ200
120847-100G
120847-25G
SIAL120847-100GEA
4840
CZK
SIAL120847-100G
SIAL120847-25G
2-Fluorobenzoyl chloride, Sigma-Aldrich®
2-Fluorobenzoyl chloride
Vzorec:
C₇H₄ClFO MW: 158.56 Bod varu: 206 Hustota: 1.32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000656 CAS číslo: 393-52-2 EINECS: 206-887-2 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...