Tisk…

2-Fluorobenzoyl chloride ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: o-Fluorobenzoyl chloride

A16096.14 A16096.22 A16096.36
A16096.14EA 1210 CZK
A16096.14 A16096.22 A16096.36
2-Fluorobenzoyl chloride ≥97%
2-Fluorobenzoyl chloride

Purity: 97% CAS No.: 393-52-2 Molecular Formula: C7H4ClFO Formula Weight: 158.56 Flash Point: 82°(179°F) Refractive Index: 1.5365 Odour: Pungent Melting Point: 4-6° Boiling Point: 207° Beilstein Reference: 636864 UN No.: 3265 MDL No.: MFCD00000656

Vzorec: C₇H₄ClFO
MW: 158,56 g/mol
Bod varu: 207 °C
Bod tání: 4…6 °C
Hustota: 1,331
Bod vzplanutí: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000656
CAS číslo: 393-52-2
EINECS: 206-887-2
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR