6-Fluoroveratraldehyde ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Fluoro-4,5-dimethoxybenzaldehyde, 4,5-Dimethoxy-2-fluorobenzaldehyde, 3,4-Dimethoxy-6-fluorobenzaldehyde
B21202.03
B21202.06
B21202.14
B21202.03EA
1490
CZK
B21202.03
B21202.06
B21202.14
6-Fluoroveratraldehyde ≥98%
6-Fluoroveratraldehyde
Purity: 98+%
CAS No.: 71924-62-4
Molecular Formula: C9H9FO3
Formula Weight: 184.17
Melting Point: 98-100°
Beilstein Reference: 2109343
Vzorec:
C₉H₉FO₃ MW: 184,17 g/mol Bod tání: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061108 CAS číslo: 71924-62-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...