2,6-Difluorobenzaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
265152-25G
265152-5G
SIAL265152-25GEA
8970
CZK
SIAL265152-25G
SIAL265152-5G
2,6-Difluorobenzaldehyde, Sigma-Aldrich®
2,6-Difluorobenzaldehyde
Vzorec:
C₇H₄F₂O Bod varu: 194 °C (1013 hPa) Bod tání: 15…17 °C Hustota: 1,31 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010293 CAS číslo: 437-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...