3-(Trifluoromethoxy)phenylboronic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(3-(Trifluoromethoxy)phenyl)metaboric acid, (3-(Trifluoromethoxy)phenyl)boronic acid, 3-(Trifluoromethoxy)benzeneboronic acid, Dihydroxy(3-(trifluoromethoxy)phenyl)borane
L19775.03
L19775.06
L19775.03EA
1450
CZK
L19775.03
L19775.06
3-(Trifluoromethoxy)phenylboronic acid ≥98%
3-(Trifluoromethoxy)phenylboronic acid
Purity: 98%
CAS No.: 179113-90-7
Molecular Formula: C7H6BF3O3
Formula Weight: 205.93
Melting Point: ca 80°
Beilstein Reference: 7531456
Vzorec:
C₇H₆BF₃O₃ MW: 205.93 Bod tání: ∼ 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD01320697 CAS číslo: 179113-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...