Pentafluorophenylboronic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2,3,4,5,6-Pentafluorobenzeneboronic acid, Dihydroxy(pentafluorophenyl)borane, (Pentafluorophenyl)metaboric acid, Pentafluorobenzeneboronic acid
B22922.06
B22922.14
B22922.06EA
3510
CZK
B22922.06
B22922.14
Pentafluorophenylboronic acid ≥97%
Pentafluorophenylboronic acid
Purity: 97%
CAS No.: 1582-24-7
Molecular Formula: C6H2BF5O2
Formula Weight: 211.88
Melting Point: 250-255°
Beilstein Reference: 3033960
Vzorec:
C₆H₂BF₅O₂ MW: 211.88 Bod tání: 250…255 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074663 CAS číslo: 1582-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...