3-Fluorophenylboronic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
3-Fluorobenzeneboronic acid, (3-Fluorophenyl)boronic acid, Dihydroxy(3-fluorophenyl)borane, (3-Fluorophenyl)metaboric acid
M-Clarity™ quality level = MQ100
441643-10G
441643-1G
SIAL441643-10GEA
11400
CZK
SIAL441643-10G
SIAL441643-1G
3-Fluorophenylboronic acid, Sigma-Aldrich®
3-Fluorophenylboronic acid
Vzorec:
C₆H₆BFO₂ MW: 139,92 g/mol Bod tání: 216…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236042 CAS číslo: 768-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...