Tisk…

2-Fluorophenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dihydroxy(2-fluorophenyl)borane, 2-Fluorobenzeneboronic acid, (2-Fluorophenyl)metaboric acid, (2-Fluorophenyl)boronic acid

M-Clarity™ quality level = MQ100

445223-10G 445223-1G
SIAL445223-10GEA 14330 CZK
SIAL445223-10G SIAL445223-1G
2-Fluorophenylboronic acid, Sigma-Aldrich®
2-Fluorophenylboronic acid
Vzorec: C₆H₆BFO₂
MW: 139.92
MDL Number: MFCD00674013
CAS číslo: 1993-03-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR