2,5-Difluorophenylboronic acid 96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(2,5-Difluorophenyl)boronic acid, 2,5-Difluorobenzeneboronic acid, (2,5-Difluorophenyl)metaboric acid, Dihydroxy(2,5-difluorophenyl)borane
B24113.03
B24113.06
B24113.14
B24113.03EA
824
CZK
B24113.03
B24113.06
B24113.14
2,5-Difluorophenylboronic acid 96%
2,5-Difluorophenylboronic acid
Purity: 96%
CAS No.: 193353-34-3
Molecular Formula: C6H5BF2O2
Formula Weight: 157.91
Beilstein Reference: 8833254
MDL No.: MFCD01863171
Vzorec:
C₆H₅BF₂O₂ MW: 157,91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01863171 CAS číslo: 193353-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...