Tisk…

1,3,5-Trifluorobenzene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

196851-25G 196851-5G
SIAL196851-25GEA 10780 CZK
SIAL196851-25G SIAL196851-5G
1,3,5-Trifluorobenzene, Sigma-Aldrich®
1,3,5-Trifluorobenzene
Vzorec: C₆H₃F₃
MW: 132,09 g/mol
Bod varu: 75 °C (1013 hPa)
Hustota: 1,277 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000333
CAS číslo: 372-38-3
EINECS: 206-751-2
UN: 2387
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR