1,2-Difluorobenzene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
126152-10G
126152-50G
SIAL126152-10GEA
1740
CZK
SIAL126152-10G
SIAL126152-50G
1,2-Difluorobenzene, Sigma-Aldrich®
1,2-Difluorobenzene
Vzorec:
C₆H₄F₂ MW: 114,09 g/mol Bod varu: 92 °C (1013 hPa) Bod tání: –34 °C Hustota: 1,15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000284 CAS číslo: 367-11-3 EINECS: 206-680-7 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...