Diphenyl phthalate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Phthalic acid diphenyl ester, Phenyl phthalate
A14449.14
A14449.22
A14449.14EA
1220
CZK
A14449.14
A14449.22
Diphenyl phthalate ≥98%
Diphenyl phthalate
Purity: 98%
CAS No.: 84-62-8
Molecular Formula: C20H14O4
Formula Weight: 318.33
Flash Point: 224°(435°F)
Odour: Odourless
Boiling Point: 400-405°
Beilstein Reference: 2473390
Merck Reference: 14,7306
MDL No.: MFCD00003038
Vzorec:
C₂₀H₁₄O₄ Bod varu: 400…405 °C Bod tání: 74…76 °C Hustota: 1,280 Bod vzplanutí: 224 °C (435 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003038 CAS číslo: 84-62-8 EINECS: 201-546-4 Merck Index: 14,7306 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...