Tisk…

Phenyl benzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Benzoic acid phenyl ester

M-Clarity™ quality level = MQ100

142719-100G 142719-5G
SIAL142719-100GEA 1750 CZK
SIAL142719-100G SIAL142719-5G
Phenyl benzoate, Sigma-Aldrich®
Phenyl benzoate
Vzorec: C₁₃H₁₀O₂
MW: 198.22
Bod varu: 299
Bod tání: 66…69 °C
Hustota: 1.041
Storage Temperature: Ambient
MDL Number: MFCD00003072
CAS číslo: 93-99-2
EINECS: 202-293-2
Merck Index: 12,07429

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR