Pyromellitic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
1,2,4,5-Benzenetetracarboxylic acid, Benzene-1,2,4,5-tetracarboxylic acid
M-Clarity™ quality level = MQ100
B4007-100G-A
B4007-500G-A
SIALB4007-100G-AEA
1290
CZK
SIALB4007-100G-A
SIALB4007-500G-A
Pyromellitic acid, Sigma-Aldrich®
Pyromellitic acid
Vzorec:
C₁₀H₆O₈ MW: 254.15 Bod varu: 516 Bod tání: 272 Hustota: 1.79 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002471 CAS číslo: 89-05-4 EINECS: 201-879-5 Merck Index: 12,08188 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...