Trimesic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Benzene-1,3,5-tricarboxylic acid, 1,3,5-Benzenetricarboxylic acid
M-Clarity™ quality level = MQ100
482749-100G
482749-500G
SIAL482749-100GEA
3450
CZK
SIAL482749-100G
SIAL482749-500G
Trimesic acid, Sigma-Aldrich®
Trimesic acid
Vzorec:
C₉H₆O₆ MW: 210,14 g/mol Bod tání: 345…350 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002517 CAS číslo: 554-95-0 EINECS: 209-077-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...