Tisk…

Thiomerzal, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Ethylmercurithiosalicylic acid sodium salt, Sodium ethylmercurithiosalicylate, Thimerosal, Merthiolat, 2-[(Ethylmercury)thio]benzoic acid sodium salt, 2-(Ethylmercuriomercapto)benzoic acid sodium salt, 2-(Ethylmercuriothio)benzoic acid sodium salt

M-Clarity™ quality level = MQ200

T5125-10G T5125-100G T5125-25G T5125-500G
SIALT5125-10GEA 9940 CZK
SIALT5125-10G SIALT5125-100G SIALT5125-25G SIALT5125-500G
Thiomerzal, Sigma-Aldrich®
Thiomerzal
Vzorec: 2-(C₂H₅HgS)C₆H₄CO₂Na
MW: 404,82 g/mol
Bod tání: 232…233 °C
Bod vzplanutí: 250 °C (ot. pohárek)
Storage Temperature: Ambient
MDL Number: MFCD00013062
CAS číslo: 54-64-8
EINECS: 200-210-4
UN: 2025
ADR: 6.1,II
Merck Index: 12,09451
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=621fb9ecf6f0ee8129acf49c938a6167645828da&vid=a4d9481c20c7c69ffe3a642fbe6cae8c3dfb117f&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR