4-(Methylthio)benzoic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
B25706.03
B25706.06
B25706.14
B25706.03EA
1370
CZK
B25706.03
B25706.06
B25706.14
4-(Methylthio)benzoic acid ≥97%
4-(Methylthio)benzoic acid
Purity: 97%
CAS No.: 13205-48-6
Molecular Formula: C8H8O2S
Formula Weight: 168.21
Melting Point: 193-196°
Beilstein Reference: 2082809
Vzorec:
C₈H₈O₂S MW: 168,22 g/mol Bod tání: 193…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002555 CAS číslo: 13205-48-6 EINECS: 236-171-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...