Tisk…

3,5-Dihydroxybenzoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: alpha-Resorcylic acid

M-Clarity™ quality level = MQ100

D110000-100G D110000-1KG
SIALD110000-100GEA 1010 CZK
SIALD110000-100G SIALD110000-1KG
3,5-Dihydroxybenzoic acid, Sigma-Aldrich®
3,5-Dihydroxybenzoic acid
Vzorec: C₇H₆O₄
Bod tání: 236…238 °C
Bod vzplanutí: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002512
CAS číslo: 99-10-5
EINECS: 202-730-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR