Tisk…

2,5-Dihydroxybenzoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,5-Dihydroxybenzoic Acid, DHB, Hydroquinonecarboxylic acid, Gentisic acid, 2,5-DHBA

M-Clarity™ quality level = MQ200

149357-100G 149357-10G 149357-25G
SIAL149357-100GEA 6220 CZK
SIAL149357-100G SIAL149357-10G SIAL149357-25G
2,5-Dihydroxybenzoic acid, Sigma-Aldrich®
2,5-Dihydroxybenzoic acid
Vzorec: C₇H₆O₄
MW: 154.12
Bod varu: 339
Bod tání: 202…206 °C
Hustota: 1.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002460
CAS číslo: 490-79-9
EINECS: 207-718-5
Merck Index: 13,04409

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR