Tisk…

2,4-Dihydroxybenzoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: beta-Resorcylic acid

M-Clarity™ quality level = MQ100

D109401-100G D109401-500G D109401-5G
SIALD109401-100GEA 1330 CZK
SIALD109401-100G SIALD109401-500G SIALD109401-5G
2,4-Dihydroxybenzoic acid, Sigma-Aldrich®
2,4-Dihydroxybenzoic acid
Vzorec: C₇H₆O₄
Bod varu: 390 °C (1013 hPa)
Bod tání: 218…220 °C
Hustota: 1,54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002451
CAS číslo: 89-86-1
EINECS: 201-946-9
Merck Index: 12,08325

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR