Tisk…

2,4,5-Trimethoxybenzoic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A19324.09 A19324.18
A19324.09EA 903 CZK
A19324.09 A19324.18
2,4,5-Trimethoxybenzoic acid ≥99%
2,4,5-Trimethoxybenzoic acid

Purity: 99% CAS No.: 490-64-2 Molecular Formula: C10H12O5 Formula Weight: 212.20 Melting Point: 143-145° MDL No.: MFCD00002435

Vzorec: C₁₀H₁₂O₅
Bod varu: 300 °C (1013 hPa)
Bod tání: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00002435
CAS číslo: 490-64-2
EINECS: 207-715-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR